Copper(II) acetate monohydrate 98%

Supplier: APOLLO SCIENTIFIC
Warning

IN1533-100G IN1533-50G
77401-400EA 48.9 USD
77401-400 77401-402
Copper(II) acetate monohydrate 98%
Copper(II) acetate monohydrate

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

Formula: (H₃CCOO)₂Cu·H₂O
Melting Pt: 115 °C
Density: 1.882 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR