1-Bromo-2-fluoro-3-iodo-5-(trifluoromethyl)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC99601-1G PC99601-25G PC99601-5G
77418-894EA 496.16 USD
77418-894 77418-896 77418-898
1-Bromo-2-fluoro-3-iodo-5-(trifluoromethyl)benzene
1-Bromo-2-fluoro-3-iodo-5-(trifluoromethyl)benzene
Formula: C₇H₂BrF₄I
MW: 368.9 g/mol
CAS Number: 2384056-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR