Methyl 4-(difluoromethoxy)-2-fluorobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

PC99579-1G PC99579-5G
77418-880EA 544.04 USD
77418-880 77418-882
Methyl 4-(difluoromethoxy)-2-fluorobenzoate
Methyl 4-(difluoromethoxy)-2-fluorobenzoate
Formula: C₉H₇F₃O₃
MW: 220.14 g/mol
MDL Number: MFCD18399137
CAS Number: 1261835-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR