2'-Fluoro-3'-iodophenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC99543-1G PC99543-5G
77418-862EA 496.16 USD
77418-862 77418-864
2'-Fluoro-3'-iodophenylacetic acid
2'-Fluoro-3'-iodophenylacetic acid
Formula: C₈H₆FIO₂
MDL Number: MFCD18396990
CAS Number: 1261675-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR