Potassium trifluoro(trifluoromethyl)borate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium (trifluoromethyl)trifluoroborate

PC99533-25G PC99533-5G
77418-858EA 1072.82 USD
77418-858 77418-860
Potassium trifluoro(trifluoromethyl)borate
Potassium trifluoro(trifluoromethyl)borate
Formula: CHBF₆
MW: 175.91 g/mol
MDL Number: MFCD09038544
CAS Number: 42298-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR