Methyl 2-difluoromethoxy-5-bromo-3-pyridinecarboxylate

Supplier: APOLLO SCIENTIFIC
Warning

PC99527-1G PC99527-5G
77418-854EA 544.04 USD
77418-854 77418-856
Methyl 2-difluoromethoxy-5-bromo-3-pyridinecarboxylate
Methyl 2-difluoromethoxy-5-bromo-3-pyridinecarboxylate
Formula: C₈H₆BrF₂NO₃
MW: 282.04 g/mol
MDL Number: MFCD34577028
CAS Number: 2559710-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR