5-Amino-2,2-difluoro-1,3-benzodioxole 94%

Supplier: APOLLO SCIENTIFIC
Danger

PC99476-10G PC99476-1G PC99476-5G
77418-834EA 28.32 USD
77418-834 77418-836 77418-838
5-Amino-2,2-difluoro-1,3-benzodioxole 94%
5-Amino-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₅F₂NO₂
Boiling Pt: 95…97 °C
Density: 1.41
MDL Number: MFCD00190144
CAS Number: 1544-85-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR