2,4,5-Trifluorobenzonitrile 94%

Supplier: APOLLO SCIENTIFIC
Warning

PC99475-100G PC99475-25G PC99475-5G
77418-828EA 172.92 USD
77418-828 77418-830 77418-832
2,4,5-Trifluorobenzonitrile 94%
2,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 168…170 °C
Density: 1.373
Flash Pt: 83 °C
MDL Number: MFCD00013289
CAS Number: 98349-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR