2,5-Difluoro-4-iodobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC99468-1G PC99468-250MG
77418-820EA 544.04 USD
77418-820 77418-822
2,5-Difluoro-4-iodobenzaldehyde
2,5-Difluoro-4-iodobenzaldehyde
Formula: C₇H₃F₂IO
MW: 268 g/mol
MDL Number: MFCD28789184
CAS Number: 1803822-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR