Ethyl 4-amino-6-chloro-5-fluoronicotinate 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC99415-10G PC99415-1G PC99415-5G
77418-802EA 832.38 USD
77418-802 77418-804 77418-806
Ethyl 4-amino-6-chloro-5-fluoronicotinate 99%
Ethyl 4-amino-6-chloro-5-fluoronicotinate
Formula: C₈H₈ClFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD34471315
CAS Number: 2454397-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR