2-(Trifluoroacetyl)thiazole monohydrate

Supplier: APOLLO SCIENTIFIC
Warning

PC99407-1G PC99407-250MG PC99407-5G
77418-796EA 234.32 USD
77418-796 77418-798 77418-800
2-(Trifluoroacetyl)thiazole monohydrate
2-(Trifluoroacetyl)thiazole monohydrate
Formula: C₅H₂F₃NOS·H₂O
MW: 199.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30742687
CAS Number: 2097800-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR