4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC99136-1G PC99136-250MG PC99136-5G
77418-772EA 578.68 USD
77418-772 77418-774 77418-776
4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₄Br₂F₄
MDL Number: MFCD33404156
CAS Number: 2090979-68-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR