Methyl 5-bromo-3-chloro-2-fluorobenzoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC99038-1G PC99038-250MG
77418-762EA 385.12 USD
77418-762 77418-764
Methyl 5-bromo-3-chloro-2-fluorobenzoate 97%
Methyl 5-bromo-3-chloro-2-fluorobenzoate
Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107276
CAS Number: 1535973-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR