3-(Difluoromethyl)-4-fluoroaniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC912796-250MG PC912796-100MG PC912796-1G
77418-594EA 182.36 USD
77418-594 77418-590 77418-592
3-(Difluoromethyl)-4-fluoroaniline 95%
3-(Difluoromethyl)-4-fluoroaniline
Formula: C₇H₆F₃N
MW: 161.1245 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11042620
CAS Number: 445303-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR