2-(Difluoromethyl)-1-fluoro-4-nitrobenzene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC912795-1G PC912795-25G PC912795-5G
77418-584EA 50.94 USD
77418-584 77418-586 77418-588
2-(Difluoromethyl)-1-fluoro-4-nitrobenzene 95%
2-(Difluoromethyl)-1-fluoro-4-nitrobenzene
Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
MDL Number: MFCD14698657
CAS Number: 63878-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR