5-Fluoro-4-iodopyridin-2-amine 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC912273-10G PC912273-1G PC912273-5G
77418-578EA 553.22 USD
77418-578 77418-580 77418-582
5-Fluoro-4-iodopyridin-2-amine 98%
5-Fluoro-4-iodopyridin-2-amine
Formula: C₅H₄FIN₂
MDL Number: MFCD28722380
CAS Number: 1649470-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR