2,3,6-Trifluorophenol 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7817D-100G
PC7817D-10G
PC7817D-25G
77418-210EA
503.3
USD
77418-210
77418-212
77418-214
2,3,6-Trifluorophenol 98%
2,3,6-Trifluorophenol
Formula:
C₆H₃F₃O Melting Pt: 31…37 °C Flash Pt: 64 °C |
MDL Number:
MFCD00061214 CAS Number: 113798-74-6 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...