Triisopropylsilyl trifluoromethanesulfonate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate

PC58047-25G PC58047-5G
77417-104EA 146.7 USD
77417-104 77417-106
Triisopropylsilyl trifluoromethanesulfonate 97%
Triisopropylsilyl trifluoromethanesulphonate
Formula: C₁₀H₂₁F₃O₃SSi
MW: 306.42 g/mol
Boiling Pt: 45…46 °C/0.03mm
Density: 1.136
Flash Pt: 100 °C
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR