2-Bromo-5-fluoro-3-(trifluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(bromomethyl)-5-fluoro-3-(trifluoromethyl)benzene

PC57804-1G PC57804-250MG PC57804-5G
77417-064EA 418.74 USD
77417-064 77417-066 77417-068
2-Bromo-5-fluoro-3-(trifluoromethyl)benzyl bromide 95%
2-Bromo-5-fluoro-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₄Br₂F₄
MW: 335.92 g/mol
MDL Number: MFCD32903689
CAS Number: 2092564-80-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR