2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57803-1G PC57803-250MG PC57803-5G
77417-058EA 392.24 USD
77417-058 77417-060 77417-062
2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile 95%
2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₂BrF₄N
MW: 268.01 g/mol
MDL Number: MFCD32903690
CAS Number: 2090965-03-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR