2-Bromo-5-(difluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57773-1G PC57773-250MG PC57773-5G
77417-048EA 544.04 USD
77417-048 77417-050 77417-052
2-Bromo-5-(difluoromethyl)benzyl bromide 95%
2-Bromo-5-(difluoromethyl)benzyl bromide
Formula: C₈H₆Br₂F₂
MDL Number: MFCD18391069
CAS Number: 1261441-93-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR