4-Bromo-2-(difluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57772-1G PC57772-250MG PC57772-5G
77417-042EA 536.92 USD
77417-042 77417-044 77417-046
4-Bromo-2-(difluoromethyl)benzyl bromide 95%
4-Bromo-2-(difluoromethyl)benzyl bromide
Formula: C₈H₆Br₂F₂
MDL Number: MFCD18391074
CAS Number: 1261448-03-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR