2-Bromo-5-(difluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57764-1G PC57764-250MG PC57764-5G
77417-034EA 536.92 USD
77417-034 77417-036 77417-038
2-Bromo-5-(difluoromethyl)benzonitrile 95%
2-Bromo-5-(difluoromethyl)benzonitrile
Formula: C₈H₄BrF₂N
MDL Number: MFCD18391059
CAS Number: 1261441-87-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR