4-Bromo-2-(difluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57763-1G PC57763-250MG PC57763-5G
77417-028EA 496.16 USD
77417-028 77417-030 77417-032
4-Bromo-2-(difluoromethyl)benzonitrile 95%
4-Bromo-2-(difluoromethyl)benzonitrile
Formula: C₈H₄BrF₂N
MDL Number: MFCD18391064
CAS Number: 1261760-22-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR