2'-Bromo-4'-chloro-5'-fluoroacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

PC57502-1G PC57502-5G
77417-016EA 578.68 USD
77417-016 77417-018
2'-Bromo-4'-chloro-5'-fluoroacetophenone
2'-Bromo-4'-chloro-5'-fluoroacetophenone
Formula: C₈H₅BrClFO
MDL Number: MFCD27943133
CAS Number: 129825-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR