2-Fluoropyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC57491-50G PC57491-100G PC57491-250G
77417-014EA 24.46 USD
77417-014 77417-010 77417-012
2-Fluoropyridine 97%
2-Fluoropyridine

JOC., 53, 2740 (1998): A useful precursor to a large range of 2,3-disubstituted pyridines. Fluorine is activated to nucleophilic displacement & C-3 metallation is possible.

Formula: C₅H₄FN
Boiling Pt: 124…126 °C
Density: 1.128
Flash Pt: 24 °C
MDL Number: MFCD00006224
CAS Number: 372-48-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR