3,5-Difluoro-4-(trifluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57489-1G PC57489-5G
77417-006EA 496.16 USD
77417-006 77417-008
3,5-Difluoro-4-(trifluoromethyl)benzonitrile 95%
3,5-Difluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₂F₅N
MDL Number: MFCD01631623
CAS Number: 261945-30-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR