2-Bromo-4-chloro-6-fluorobenzoyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57435-1G PC57435-250MG PC57435-5G
77416-996EA 496.16 USD
77416-996 77416-998 77417-000
2-Bromo-4-chloro-6-fluorobenzoyl chloride 95%
2-Bromo-4-chloro-6-fluorobenzoyl chloride
Formula: C₇H₂BrCl₂FO
MW: 271.9 g/mol
MDL Number: MFCD28733936
CAS Number: 1805575-89-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR