Methyl 4-cyano-5-fluoro-2-methylbenzoate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57432-5G PC57432-1G PC57432-250MG
77416-994EA 2058 USD
77416-994 77416-990 77416-992
Methyl 4-cyano-5-fluoro-2-methylbenzoate 95%
Methyl 4-cyano-5-fluoro-2-methylbenzoate
Formula: C₁₀H₈FNO₂
MDL Number: MFCD23707556
CAS Number: 1427377-85-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR