1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57411-1G PC57411-250MG PC57411-5G
77416-980EA 536.92 USD
77416-980 77416-982 77416-984
1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene 95%
1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene
Formula: C₆HBrCl₂FI
MDL Number: MFCD34166239
CAS Number: 2676196-06-6
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR