2-Bromo-4-chloro-5-fluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC57125-1G PC57125-250MG PC57125-5G
77416-970EA 285.26 USD
77416-970 77416-972 77416-974
2-Bromo-4-chloro-5-fluorobenzonitrile 95%
2-Bromo-4-chloro-5-fluorobenzonitrile
Formula: C₇H₂BrClFN
MDL Number: MFCD30184437
CAS Number: 1893432-29-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR