2-Bromo-4-(trifluoromethoxy)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(bromomethyl)-4-(trifluoromethoxy)benzene

PC57117-1G PC57117-250MG PC57117-5G
77416-964EA 365.76 USD
77416-964 77416-966 77416-968
2-Bromo-4-(trifluoromethoxy)benzyl bromide 95%
2-Bromo-4-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅Br₂F₃O
MW: 333.93 g/mol
MDL Number: MFCD32903683
CAS Number: 2168988-02-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR