3-(1,1-Difluoroethyl)phenacyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-[3-(1,1-difluoroethyl)phenyl]ethanone

PC56998-1G PC56998-250MG PC56998-5G
77416-938EA 496.16 USD
77416-938 77416-940 77416-942
3-(1,1-Difluoroethyl)phenacyl bromide 95%
3-(1,1-Difluoroethyl)phenacyl bromide
Formula: C₁₀H₉BrF₂O
MW: 263.08 g/mol
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR