5-Chloro-4-fluoro-2-(trifluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-5-chloro-4-fluoro-2-(trifluoromethyl)benzene

PC56989-1G PC56989-250MG PC56989-5G
77416-930EA 578.68 USD
77416-930 77416-932 77416-934
5-Chloro-4-fluoro-2-(trifluoromethyl)benzyl bromide 95%
5-Chloro-4-fluoro-2-(trifluoromethyl)benzyl bromide
Formula: C₈H₄BrClF₄
MW: 291.47 g/mol
MDL Number: MFCD31696997
CAS Number: 2090498-26-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR