5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC56987-1G PC56987-250MG PC56987-5G
77416-924EA 496.16 USD
77416-924 77416-926 77416-928
5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile 95%
5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₂ClF₄N
MDL Number: MFCD28752675
CAS Number: 1807257-71-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR