Calcium hexafluorophosphate

Supplier: APOLLO SCIENTIFIC
Danger

PC53079-1G PC53079-25G PC53079-5G
77416-614EA 75.4 USD
77416-614 77416-616 77416-618
Calcium hexafluorophosphate
Calcium hexafluorophosphate
Formula: Ca₂PF₆
MW: 330.01 g/mol
MDL Number: MFCD28963758
CAS Number: 78415-39-1
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR