2-Bromo-5-chloro-4-fluorobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Bromo-5-chloro-4-fluorophenyl)methanol

PC50331-1G PC50331-5G
77416-362EA 350.48 USD
77416-362 77416-364
2-Bromo-5-chloro-4-fluorobenzyl alcohol
2-Bromo-5-chloro-4-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
MW: 239.47 g/mol
MDL Number: MFCD30297360
CAS Number: 1067882-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR