2-Bromo-5-chloro-4-fluorobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC50306-1G PC50306-5G
77416-358EA 578.68 USD
77416-358 77416-360
2-Bromo-5-chloro-4-fluorobenzyl bromide
2-Bromo-5-chloro-4-fluorobenzyl bromide
Formula: C₇H₄Br₂ClF
MDL Number: MFCD30296109
CAS Number: 1067882-52-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR