7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC502948-1G PC502948-250MG
77416-200EA 1210.36 USD
77416-200 77416-202
7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine 95%
7-Bromo-3-(difluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₄BrF₂N₃
MDL Number: MFCD31922795
CAS Number: 2331325-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR