7-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC502937-1G PC502937-250MG
77416-196EA 942.4 USD
77416-196 77416-198
7-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
7-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₃BrF₃N₃
MW: 266.02 g/mol
MDL Number: MFCD15144602
CAS Number: 1021923-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR