2,3-Difluoro-6-(trifluoromethoxy)benzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC502499-1G PC502499-5G
77416-190EA 496.16 USD
77416-190 77416-192
2,3-Difluoro-6-(trifluoromethoxy)benzonitrile
2,3-Difluoro-6-(trifluoromethoxy)benzonitrile
Formula: C₈H₂F₅NO
MW: 223.1 g/mol
MDL Number: MFCD28791002
CAS Number: 1803828-42-3
UN: 3276

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR