2,6-Difluoro-3-(trifluoromethoxy)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC502468-1G PC502468-250MG PC502468-5G
77416-178EA 496.16 USD
77416-178 77416-180 77416-182
2,6-Difluoro-3-(trifluoromethoxy)benzonitrile 95%
2,6-Difluoro-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₂F₅NO
MW: 223.1 g/mol
MDL Number: MFCD28791008
CAS Number: 1806290-43-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR