2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC502457-250MG PC502457-5G PC502457-1G
77416-174EA 166.06 USD
77416-174 77416-176 77416-172
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide
Formula: C₈H₄BrF₅O
MW: 291.01 g/mol
MDL Number: MFCD31561157
CAS Number: 2244081-55-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR