1-Bromo-4,5-difluoro-2-nitrobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4,5-difluoronitrobenzene

PC502293-10G PC502293-5G
77416-168EA 36.68 USD
77416-168 77416-170
1-Bromo-4,5-difluoro-2-nitrobenzene 98%
1-Bromo-4,5-difluoro-2-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.98 g/mol
Boiling Pt: 225.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06808551
CAS Number: 321-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR