2,6-Dichloro-4-fluorobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC502275-1G PC502275-250MG
77416-164EA 578.68 USD
77416-164 77416-166
2,6-Dichloro-4-fluorobenzyl bromide 95%
2,6-Dichloro-4-fluorobenzyl bromide
Formula: C₇H₄BrCl₂F
Storage Temperature: Refrigerator
MDL Number: MFCD28346557
CAS Number: 1803784-34-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR