4-Bromo-5-fluoro-2-methoxybenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC501858-1G PC501858-5G
77416-146EA 496.16 USD
77416-146 77416-148
4-Bromo-5-fluoro-2-methoxybenzonitrile
4-Bromo-5-fluoro-2-methoxybenzonitrile
Formula: C₈H₅BrFNO
MDL Number: MFCD23707399
CAS Number: 1427360-03-9
UN: 3439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR