2-Bromo-3,6-difluorophenacyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC501817-1G PC501817-250MG PC501817-5G
77416-140EA 619.44 USD
77416-140 77416-142 77416-144
2-Bromo-3,6-difluorophenacyl bromide 95%
2-Bromo-3,6-difluorophenacyl bromide
Formula: C₈H₄Br₂F₂O
MDL Number: MFCD28061331
CAS Number: 1807197-48-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR