2-Bromo-3,6-difluorobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC501813-1G PC501813-250MG PC501813-5G
77416-134EA 578.68 USD
77416-134 77416-136 77416-138
2-Bromo-3,6-difluorobenzyl bromide 95%
2-Bromo-3,6-difluorobenzyl bromide
Formula: C₇H₄Br₂F₂
MW: 285.91 g/mol
MDL Number: MFCD28061450
CAS Number: 1805119-92-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR