3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC501730-1G PC501730-250MG
77416-130EA 605.18 USD
77416-130 77416-132
3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride 95%
3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₃ClF₃NO₅S
MDL Number: MFCD28801862
CAS Number: 1351094-07-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR