2-Fluoro-6-nitrobenzenesulphonamide

Supplier: APOLLO SCIENTIFIC
Warning

PC501663-1G PC501663-250MG PC501663-5G
77416-122EA 804.86 USD
77416-122 77416-124 77416-126
2-Fluoro-6-nitrobenzenesulphonamide
2-Fluoro-6-nitrobenzenesulphonamide
Formula: C₆H₅FN₂O₄S
MDL Number: MFCD12912984
CAS Number: 1210309-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR